siRNA Details
| Gene Symbol: OR5M3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004742.2 | NP_001004742.2 | 1 | 465 | ||
| GenBank mRNA | BC136982.1 | - | 1 | 465 | 9020974 | |
| BC136983.1 | - | 1 | 465 | 9020975 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5M3| Gene Symbol: OR5M3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004742.2 | NP_001004742.2 | 1 | 465 | ||
| GenBank mRNA | BC136982.1 | - | 1 | 465 | 9020974 | |
| BC136983.1 | - | 1 | 465 | 9020975 | ||