siRNA Details
| Gene Symbol: LRRC55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005210.2 | NP_001005210.1 | 2 | 1537 | ||
| GenBank mRNA | AK127591.1 | - | 2 | 1537 | ||
| BC136737.1 | - | 2 | 1495 | 9020727 | ||
| BC136739.1 | - | 2 | 1495 | 9020729 | ||

LRRC55
Genomic Map