siRNA Details
| Gene Symbol: EPRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004446.2 | NP_004437.2 | 28 | 4298 | ||
| XM_017000614.1 | XP_016856103.1 | 17 | 2554 | |||
| GenBank mRNA | AK298192.1 | - | 9 | 1122 | ||
| AY493416.1 | - | 26 | 3845 | |||
| BC126275.1 | - | 28 | 4166 | 8991991 | ||
| BC136465.1 | - | 28 | 4066 | 9020454 | ||
| CR933648.1 | - | 28 | 4138 | |||
| X54326.1 | - | 26 | 3870 | |||

EPRS
Genomic Map