siRNA Details
| Gene Symbol: C10orf107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173554.2 | NP_775825.1 | 5 | 700 | ||
| XM_005269598.3 | XP_005269655.1 | 6 | 690 | |||
| XM_005269599.4 | XP_005269656.1 | 6 | 798 | |||
| XM_005269600.4 | XP_005269657.1 | 5 | 601 | |||
| XM_006717690.3 | XP_006717753.1 | 6 | 672 | |||
| GenBank mRNA | BC041932.1 | - | 5 | 715 | 5300631 | |

C10orf107
Genomic Map