siRNA Details
| Gene Symbol: ZCCHC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK024784.1 | - | 1 | 487 | ||
| AK075279.1 | - | 4 | 1596 | |||
| BC028617.1 | - | 4 | 1746 | 4821347 | ||
| BC039249.1 | - | 4 | 1738 | |||
| BC053564.1 | - | 4 | 1643 | 6193781 | ||
| BX648607.1 | - | 1 | 630 | |||

ZCCHC24
Genomic Map