siRNA Details
| Gene Symbol: ZCCHC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153367.3 | NP_699198.2 | 3 | 782 | ||
| XM_011539452.2 | XP_011537754.1 | 3 | 733 | |||
| GenBank mRNA | AK075279.1 | - | 3 | 634 | ||
| AK293361.1 | - | 3 | 421 | |||
| AK303734.1 | - | 3 | 413 | |||
| AK308876.1 | - | 4 | 713 | |||
| BC028617.1 | - | 3 | 784 | 4821347 | ||
| BC039249.1 | - | 3 | 777 | |||
| BC053564.1 | - | 3 | 681 | 6193781 | ||

ZCCHC24
Genomic Map