siRNA Details
| Gene Symbol: C11orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256088.1 | NP_001243017.1 | 4 | 1255 | ||
| NM_145013.2 | NP_659450.1 | 4 | 1244 | |||
| NR_045767.1 | 4 | 1178 | ||||
| XM_011542654.2 | XP_011540956.1 | 4 | 1174 | |||
| XM_011542655.2 | XP_011540957.1 | 4 | 1136 | |||
| GenBank mRNA | AK125634.1 | - | 4 | 1152 | ||
| BC025756.1 | - | 4 | 1244 | 5201867 | ||

C11orf45
Genomic Map