siRNA Details
| Gene Symbol: HYLS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134793.1 | NP_001128265.1 | 3 | 1165 | ||
| NM_145014.2 | NP_659451.1 | 4 | 918 | |||
| XM_005271430.2 | XP_005271487.1 | 2 | 1115 | |||
| XM_006718777.3 | XP_006718840.1 | 2 | 432 | |||
| XM_011542659.2 | XP_011540961.1 | 2 | 593 | |||
| XM_017017320.1 | XP_016872809.1 | 3 | 884 | |||
| XM_017017321.1 | XP_016872810.1 | 3 | 881 | |||
| GenBank mRNA | AK057477.1 | - | 4 | 920 | ||
| AK127394.1 | - | 3 | 1168 | |||
| BC015047.1 | - | 3 | 506 | 3909690 | ||

HYLS1
Genomic Map