siRNA Details
| Gene Symbol: OR10S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004474.1 | NP_001004474.1 | 1 | 685 | ||
| GenBank mRNA | BC137037.1 | - | 1 | 775 | 9021033 | |
| BC137038.1 | - | 1 | 775 | 9021034 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR10S1| Gene Symbol: OR10S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004474.1 | NP_001004474.1 | 1 | 685 | ||
| GenBank mRNA | BC137037.1 | - | 1 | 775 | 9021033 | |
| BC137038.1 | - | 1 | 775 | 9021034 | ||