siRNA Details
| Gene Symbol: OR9Q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005283.2 | NP_001005283.1 | 1 | 577 | ||
| GenBank mRNA | BC140718.1 | - | 1 | 577 | 9021586 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR9Q2| Gene Symbol: OR9Q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005283.2 | NP_001005283.1 | 1 | 577 | ||
| GenBank mRNA | BC140718.1 | - | 1 | 577 | 9021586 | |