siRNA Details
| Gene Symbol: OR10Q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004471.2 | NP_001004471.1 | 1 | 389 | ||
| GenBank mRNA | BC140731.1 | - | 1 | 389 | 9021599 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR10Q1| Gene Symbol: OR10Q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004471.2 | NP_001004471.1 | 1 | 389 | ||
| GenBank mRNA | BC140731.1 | - | 1 | 389 | 9021599 | |