siRNA Details
| Gene Symbol: OR5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001954.1 | NP_001001954.1 | 1 | 306 | ||
| GenBank mRNA | BC136991.1 | - | 1 | 329 | 9020983 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5A2| Gene Symbol: OR5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001954.1 | NP_001001954.1 | 1 | 306 | ||
| GenBank mRNA | BC136991.1 | - | 1 | 329 | 9020983 | |