siRNA Details
| Gene Symbol: OOSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173801.4 | NP_776162.2 | 2 | 294 | ||
| GenBank mRNA | AK093517.1 | - | 2 | 160 | ||
| BC036256.1 | - | 2 | 160 | 5270384 | ||
| BC048121.1 | - | 2 | 294 | 5266642 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OOSP2| Gene Symbol: OOSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173801.4 | NP_776162.2 | 2 | 294 | ||
| GenBank mRNA | AK093517.1 | - | 2 | 160 | ||
| BC036256.1 | - | 2 | 160 | 5270384 | ||
| BC048121.1 | - | 2 | 294 | 5266642 | ||