siRNA Details
| Gene Symbol: DDIAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145018.3 | NP_659455.3 | 6 | 1950 | ||
| XM_011544835.1 | XP_011543137.1 | 5 | 1812 | |||
| XM_011544836.2 | XP_011543138.1 | 5 | 1767 | |||
| XM_011544837.1 | XP_011543139.1 | 4 | 1662 | |||
| XM_011544838.2 | XP_011543140.1 | 5 | 1796 | |||
| XM_017017352.1 | XP_016872841.1 | 4 | 1667 | |||
| XM_017017353.1 | XP_016872842.1 | 4 | 1634 | |||
| GenBank mRNA | AK023998.1 | - | 1 | 1235 | ||
| AK096157.1 | - | 6 | 1915 | |||
| AK292399.1 | - | 6 | 1909 | |||
| AK297366.1 | - | 4 | 1635 | |||
| BC039268.1 | - | 6 | 1918 | 5296465 | ||

DDIAS
Genomic Map