siRNA Details
| Gene Symbol: DDIAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145018.3 | NP_659455.3 | 4 | 479 | ||
| XM_011544835.1 | XP_011543137.1 | 3 | 341 | |||
| XM_011544836.2 | XP_011543138.1 | 3 | 296 | |||
| XM_011544837.1 | XP_011543139.1 | 2 | 191 | |||
| XM_011544838.2 | XP_011543140.1 | 4 | 443 | |||
| GenBank mRNA | AK058145.1 | - | 5 | 690 | ||
| AK096157.1 | - | 4 | 444 | |||
| AK292399.1 | - | 4 | 438 | |||
| AK299581.1 | - | 4 | 437 | |||
| BC039268.1 | - | 4 | 447 | 5296465 | ||

DDIAS
Genomic Map