siRNA Details
| Gene Symbol: SPERT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286341.1 | NP_001273270.1 | 2 | 385 | ||
| NM_001286342.1 | NP_001273271.1 | 3 | 351 | |||
| NM_152719.2 | NP_689932.1 | 3 | 466 | |||
| XM_011534971.2 | XP_011533273.1 | 4 | 1821 | |||
| GenBank mRNA | AK093129.1 | - | 3 | 459 | ||
| AK292350.1 | - | 2 | 687 | |||
| BC027609.2 | - | 2 | 285 | 4829821 | ||
| HY007057.1 | - | 3 | 458 | |||
| HY016268.1 | - | 3 | 351 | |||
| HY026582.1 | - | 2 | 269 | |||

SPERT
Genomic Map