siRNA Details
| Gene Symbol: FAM124A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242312.1 | NP_001229241.1 | 3, 4 | 1011 | ||
| NM_145019.3 | NP_659456.3 | 4, 5 | 1119 | |||
| GenBank mRNA | AK055269.1 | - | 3, 4 | 978 | ||
| AK096364.1 | - | 4, 5 | 1082 | |||
| BC051771.1 | - | 3, 4 | 926 | 5722982 | ||

FAM124A
Genomic Map