siRNA Details
| Gene Symbol: FAM181B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175885.3 | NP_787081.2 | 1 | 335 | ||
| GenBank mRNA | BC150621.1 | - | 1 | 319 | 9056991 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM181B| Gene Symbol: FAM181B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175885.3 | NP_787081.2 | 1 | 335 | ||
| GenBank mRNA | BC150621.1 | - | 1 | 319 | 9056991 | |