siRNA Details
| Gene Symbol: CCDC89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152723.2 | NP_689936.1 | 1 | 953 | ||
| GenBank mRNA | AK095478.1 | - | 1 | 953 | ||
| BC044814.1 | - | 1 | 849 | 5167843 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC89| Gene Symbol: CCDC89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152723.2 | NP_689936.1 | 1 | 953 | ||
| GenBank mRNA | AK095478.1 | - | 1 | 953 | ||
| BC044814.1 | - | 1 | 849 | 5167843 | ||