siRNA Details
Gene Symbol: RASGEF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282862.1 | NP_001269791.1 | 10 | 1258 | ||
NM_145313.3 | NP_660356.2 | 10 | 1230 | |||
XM_005271809.3 | XP_005271866.1 | 9 | 1108 | |||
XM_011539500.2 | XP_011537802.1 | 9 | 1218 | |||
GenBank mRNA | AK095136.1 | - | 10 | 1258 | ||
AK127432.1 | - | 10 | 3656 | |||
BC022548.1 | - | 10 | 1230 | 4814750 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
