siRNA Details
| Gene Symbol: RASGEF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282862.1 | NP_001269791.1 | 2, 3 | 317 | ||
| NM_145313.3 | NP_660356.2 | 2, 3 | 289 | |||
| GenBank mRNA | AK095136.1 | - | 2, 3 | 317 | ||
| AK127432.1 | - | 2, 3 | 2715 | |||
| BC022548.1 | - | 2, 3 | 289 | 4814750 | ||
| HY118234.1 | - | 2, 3 | 290 | |||

RASGEF1A
Genomic Map