siRNA Details
| Gene Symbol: FAM171A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010924.1 | NP_001010924.1 | 2 | 262 | ||
| XM_011519378.2 | XP_011517680.1 | 2 | 922 | |||
| XM_017015904.1 | XP_016871393.1 | 2 | 183 | |||
| GenBank mRNA | AL050367.1 | - | 2 | 262 | ||
| AY683003.1 | - | 2 | 309 | |||
| BC109302.1 | - | 2 | 262 | 40035211 | ||
| BC109303.1 | - | 2 | 262 | 40035213 | ||
| BX496483.1 | - | 2 | 267 | |||

FAM171A1
Genomic Map