siRNA Details
| Gene Symbol: FAM171A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010924.1 | NP_001010924.1 | 8 | 2397 | ||
| XM_011519378.2 | XP_011517680.1 | 8 | 3057 | |||
| XM_017015904.1 | XP_016871393.1 | 8 | 2318 | |||
| XM_017015905.1 | XP_016871394.1 | 6 | 1976 | |||
| GenBank mRNA | AK022946.1 | - | 1 | 1087 | ||
| AL050367.1 | - | 8 | 2396 | |||
| AY683003.1 | - | 8 | 2444 | |||
| BC034232.1 | - | 1 | 741 | |||
| BC109302.1 | - | 8 | 2397 | 40035211 | ||
| BC109303.1 | - | 8 | 2397 | 40035213 | ||

FAM171A1
Genomic Map