siRNA Details
| Gene Symbol: SPATA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166271.2 | NP_001159743.1 | 12 | 3972 | ||
| NM_001286792.1 | NP_001273721.1 | 14 | 4296 | |||
| NM_001286793.1 | NP_001273722.1 | 8 | 1520 | |||
| NM_001286794.1 | NP_001273723.1 | 10 | 1914 | |||
| NM_001286795.1 | NP_001273724.1 | 9 | 1706 | |||
| NM_153023.3 | NP_694568.1 | 11 | 2208 | |||
| GenBank mRNA | AK055770.1 | - | 11 | 2176 | ||
| AK074117.1 | - | 2 | 418 | |||
| AK092759.1 | - | 2 | 173 | |||
| AK097217.1 | - | 5 | 849 | |||
| AK123031.1 | - | 9 | 1606 | |||
| AK123225.1 | - | 1 | 817 | |||
| AK160371.1 | - | 8 | 1395 | |||
| AK297394.1 | - | 8 | 1490 | |||
| AK298717.1 | - | 10 | 1816 | |||
| AK299981.1 | - | 9 | 1649 | |||
| AK301149.1 | - | 8 | 1463 | |||
| AK307528.1 | - | 3 | 422 | |||
| BC109290.1 | - | 10 | 1918 | 40034357 | ||
| BC109291.1 | - | 10 | 1918 | 40034359 | ||
| BX648244.1 | - | 14 | 4118 | |||

SPATA13
Genomic Map