siRNA Details
| Gene Symbol: CPNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152727.5 | NP_689940.3 | 6, 7 | 875 | ||
| GenBank mRNA | AF492484.1 | - | 6, 7 | 826 | ||
| AK292269.1 | - | 5, 6 | 784 | |||
| AK315394.1 | - | 6, 7 | 778 | |||
| BC053658.1 | - | 7, 8 | 949 | 5750192 | ||
| CR749617.1 | - | 5, 6 | 536 | |||

CPNE2
Genomic Map