siRNA Details
Gene Symbol: CCDC162P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028595.1 | 3 | 426 | |||
GenBank mRNA | BC107113.2 | - | 4 | 566 | 40022743 | |
BC107114.2 | - | 3 | 426 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC162P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028595.1 | 3 | 426 | |||
GenBank mRNA | BC107113.2 | - | 4 | 566 | 40022743 | |
BC107114.2 | - | 3 | 426 |