siRNA Details
| Gene Symbol: FKBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002014.3 | NP_002005.1 | 8 | 1071 | ||
| XM_011520929.2 | XP_011519231.1 | 8 | 2064 | |||
| GenBank mRNA | AK313918.1 | - | 8 | 1036 | ||
| BC001786.1 | - | 8 | 1026 | 3542330 | ||
| BC002887.2 | - | 5 | 430 | |||
| BC007924.2 | - | 8 | 971 | 4304462 | ||
| M88279.1 | - | 8 | 972 | |||

FKBP4
Genomic Map