siRNA Details
| Gene Symbol: FAM162B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085480.2 | NP_001078949.1 | 3 | 473 | ||
| GenBank mRNA | BC038997.1 | - | 3 | 472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM162B| Gene Symbol: FAM162B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085480.2 | NP_001078949.1 | 3 | 473 | ||
| GenBank mRNA | BC038997.1 | - | 3 | 472 | ||