siRNA Details
| Gene Symbol: BEND6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152731.2 | NP_689944.2 | 4 | 1031 | ||
| XM_005248889.1 | XP_005248946.1 | 2 | 199 | |||
| XM_011514346.2 | XP_011512648.1 | 4 | 1019 | |||
| XM_017010406.1 | XP_016865895.1 | 4 | 582 | |||
| XM_017010407.1 | XP_016865896.1 | 4 | 565 | |||
| XM_017010408.1 | XP_016865897.1 | 5 | 1112 | |||
| GenBank mRNA | AK054724.1 | - | 4 | 716 | ||
| AK303705.1 | - | 3 | 401 | |||
| BC022988.1 | - | 3 | 426 | 4932760 | ||
| DC411761.1 | - | 3 | 401 | |||

BEND6
Genomic Map