siRNA Details
| Gene Symbol: ADGRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153838.3 | NP_722580.3 | 6 | 1444 | ||
| GenBank mRNA | AK095395.1 | - | 6 | 1356 | ||
| AY255556.1 | - | 2 | 814 | |||
| DQ315370.1 | - | 6 | 1431 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADGRF4| Gene Symbol: ADGRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153838.3 | NP_722580.3 | 6 | 1444 | ||
| GenBank mRNA | AK095395.1 | - | 6 | 1356 | ||
| AY255556.1 | - | 2 | 814 | |||
| DQ315370.1 | - | 6 | 1431 | |||