siRNA Details
| Gene Symbol: ADGRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153838.3 | NP_722580.3 | 3, 4 | 406 | ||
| GenBank mRNA | AK095395.1 | - | 3, 4 | 318 | ||
| DA445316.1 | - | 3, 4 | 318 | |||
| DQ315370.1 | - | 3, 4 | 393 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADGRF4| Gene Symbol: ADGRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153838.3 | NP_722580.3 | 3, 4 | 406 | ||
| GenBank mRNA | AK095395.1 | - | 3, 4 | 318 | ||
| DA445316.1 | - | 3, 4 | 318 | |||
| DQ315370.1 | - | 3, 4 | 393 | |||