siRNA Details
| Gene Symbol: C6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109938.2 | NP_001103408.1 | 4, 5 | 954 | ||
| NM_001161376.1 | NP_001154848.1 | 4, 5 | 1497 | |||
| NM_145029.3 | NP_659466.2 | 3, 4 | 552 | |||
| XM_011514384.1 | XP_011512686.1 | 5, 6 | 1046 | |||
| GenBank mRNA | AK127564.1 | - | 4, 5 | 914 | ||
| BC016167.1 | - | 4, 5 | 1105 | 3509511 | ||
| BC064397.1 | - | 4, 5 | 1115 | |||
| BM695485.1 | - | 3, 4 | 427 | |||

C6orf136
Genomic Map