siRNA Details
| Gene Symbol: RNF182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165032.1 | NP_001158504.1 | 4 | 1216 | ||
| NM_001165033.1 | NP_001158505.1 | 2 | 995 | |||
| NM_001165034.1 | NP_001158506.1 | 3 | 1103 | |||
| NM_152737.3 | NP_689950.1 | 3 | 1133 | |||
| XM_017010448.1 | XP_016865937.1 | 4 | 1153 | |||
| XM_017010449.1 | XP_016865938.1 | 3 | 1217 | |||
| XM_017010450.1 | XP_016865939.1 | 2 | 1201 | |||
| GenBank mRNA | AK090576.1 | - | 2 | 952 | ||
| AK315528.1 | - | 3 | 1114 | |||
| BC030666.1 | - | 3 | 1033 | 5259219 | ||
| BC050030.1 | - | 2 | 857 | 5297801 | ||

RNF182
Genomic Map