siRNA Details
| Gene Symbol: FAM200A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145111.3 | NP_659802.1 | 2 | 669 | ||
| XM_011515911.2 | XP_011514213.1 | 2 | 473 | |||
| XM_011515913.2 | XP_011514215.1 | 2 | 412 | |||
| GenBank mRNA | AK094113.1 | - | 2 | 581 | ||
| AK290724.1 | - | 2 | 669 | |||
| AK315452.1 | - | 2 | 605 | |||
| AL713672.1 | - | 1 | 382 | |||
| BC009536.1 | - | 2 | 446 | |||
| BC040214.1 | - | 2 | 586 | 5166971 | ||
| BC126337.1 | - | 2 | 531 | 8992053 | ||
| BC130371.1 | - | 2 | 531 | 40146402 | ||

FAM200A
Genomic Map