siRNA Details
| Gene Symbol: FAM200A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145111.3 | NP_659802.1 | 2 | 708 | ||
| XM_011515911.2 | XP_011514213.1 | 2 | 512 | |||
| XM_011515913.2 | XP_011514215.1 | 2 | 451 | |||
| GenBank mRNA | AK094113.1 | - | 2 | 620 | ||
| AK290724.1 | - | 2 | 708 | |||
| AK315452.1 | - | 2 | 644 | |||
| AL713672.1 | - | 1 | 421 | |||
| BC009536.1 | - | 2 | 485 | |||
| BC040214.1 | - | 2 | 625 | 5166971 | ||
| BC126337.1 | - | 2 | 570 | 8992053 | ||
| BC130371.1 | - | 2 | 570 | 40146402 | ||

FAM200A
Genomic Map