siRNA Details
| Gene Symbol: FOXK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037165.1 | NP_001032242.1 | 6 | 1401 | ||
| XM_011515191.2 | XP_011513493.1 | 8 | 1072 | |||
| GenBank mRNA | AK027883.1 | - | 1 | 1189 | ||
| AK122663.1 | - | 7 | 891 | |||
| AK302243.1 | - | 6 | 1040 | |||
| BC038434.1 | - | 9 | 1145 | |||

FOXK1
Genomic Map