siRNA Details
| Gene Symbol: THSD7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015204.2 | NP_056019.1 | 25 | 4799 | ||
| XM_006715659.1 | XP_006715722.1 | 25 | 4798 | |||
| XM_011515193.2 | XP_011513495.1 | 25 | 7123 | |||
| XM_011515194.1 | XP_011513496.1 | 25 | 4564 | |||
| GenBank mRNA | AB023177.2 | - | 24 | 4083 | ||
| BC016980.2 | - | 1 | 99 | |||

THSD7A
Genomic Map