siRNA Details
| Gene Symbol: RSBN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198467.2 | NP_940869.2 | 5 | 1576 | ||
| GenBank mRNA | AK127713.1 | - | 3 | 771 | ||
| BC059402.1 | - | 5 | 1547 | 30347338 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RSBN1L| Gene Symbol: RSBN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198467.2 | NP_940869.2 | 5 | 1576 | ||
| GenBank mRNA | AK127713.1 | - | 3 | 771 | ||
| BC059402.1 | - | 5 | 1547 | 30347338 | ||