siRNA Details
| Gene Symbol: FBXL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111038.1 | NP_001104508.1 | 13 | 1417 | ||
| NM_001287150.1 | NP_001274079.1 | 13 | 1417 | |||
| NM_145032.3 | NP_659469.3 | 13 | 1594 | |||
| NR_105043.1 | 14 | 1508 | ||||
| XM_005250205.3 | XP_005250262.1 | 14 | 1500 | |||
| XM_005250207.3 | XP_005250264.1 | 13 | 1401 | |||
| XM_005250208.3 | XP_005250265.1 | 14 | 1500 | |||
| XM_005250209.2 | XP_005250266.1 | 13 | 1398 | |||
| XM_006715898.2 | XP_006715961.1 | 8 | 845 | |||
| XM_011515928.2 | XP_011514230.1 | 14 | 1500 | |||
| XM_011515929.2 | XP_011514231.1 | 14 | 1500 | |||
| XM_011515930.2 | XP_011514232.1 | 13 | 1464 | |||
| XM_011515932.2 | XP_011514234.1 | 14 | 1500 | |||
| XM_017011850.1 | XP_016867339.1 | 14 | 1444 | |||
| XM_017011851.1 | XP_016867340.1 | 14 | 1500 | |||
| XM_017011852.1 | XP_016867341.1 | 14 | 1650 | |||
| XM_017011853.1 | XP_016867342.1 | 13 | 1450 | |||
| GenBank mRNA | AK097537.1 | - | 13 | 1594 | ||
| AL713709.1 | - | 4 | 316 | |||
| AY359239.1 | - | 13 | 1301 | |||
| BC020572.2 | - | 7 | 671 | |||
| BC026121.1 | - | 14 | 1511 | 4826546 | ||
| BC031285.1 | - | 13 | 1409 | 5298780 | ||

FBXL13
Genomic Map