siRNA Details
| Gene Symbol: SLC35F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029858.3 | NP_001025029.2 | 5, 6 | 993 | ||
| XM_005266865.4 | XP_005266922.1 | 5, 6 | 1293 | |||
| GenBank mRNA | AK128717.1 | - | 3, 4 | 341 | ||
| AK295860.1 | - | 5, 6 | 690 | |||
| BC028615.1 | - | 5, 6 | 996 | 4817165 | ||
| BC036493.1 | - | 5, 6 | 761 | |||
| BC114925.1 | - | 6, 7 | 867 | 40025808 | ||
| BX640761.1 | - | 2, 3 | 254 | |||

SLC35F1
Genomic Map