siRNA Details
| Gene Symbol: FAM83B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010872.2 | NP_001010872.1 | 5 | 2917 | ||
| XM_006715022.3 | XP_006715085.1 | 5 | 2946 | |||
| XM_011514394.2 | XP_011512696.1 | 5 | 2959 | |||
| XM_011514395.2 | XP_011512697.1 | 5 | 3948 | |||
| XM_017010478.1 | XP_016865967.1 | 5 | 2943 | |||
| GenBank mRNA | BC101628.1 | - | 4 | 2876 | 8069134 | |
| BC112275.1 | - | 4 | 2876 | 8327743 | ||

FAM83B
Genomic Map