siRNA Details
| Gene Symbol: TSPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010873.2 | NP_001010873.1 | 4 | 934 | ||
| NM_001159726.1 | NP_001153198.1 | 4 | 744 | |||
| XM_011514396.2 | XP_011512698.1 | 4 | 737 | |||
| XM_011514397.2 | XP_011512699.1 | 4 | 1266 | |||

TSPO2
Genomic Map