siRNA Details
| Gene Symbol: UNC5CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173561.2 | NP_775832.2 | 6 | 1177 | ||
| GenBank mRNA | AY510109.1 | - | 5 | 1078 | ||
| BC035284.1 | - | 6 | 1167 | 5189247 | ||
| BM312066.1 | - | 3 | 219 | |||
| JF681947.1 | - | 6 | 1195 | |||

UNC5CL
Genomic Map