siRNA Details
| Gene Symbol: PXT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152990.3 | NP_694535.2 | 4, 5 | 779 | ||
| XM_011514400.2 | XP_011512702.1 | 3, 4 | 452 | |||
| GenBank mRNA | AF486827.1 | - | 1, 2 | 76 | ||
| BC031105.1 | - | 4, 5 | 780 | 5297409 | ||
| BC107049.2 | - | 1, 2 | 66 | 40003975 | ||

PXT1
Genomic Map