siRNA Details
| Gene Symbol: TMEM130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134450.1 | NP_001127922.1 | 7 | 1234 | ||
| NM_001134451.1 | NP_001127923.1 | 6 | 928 | |||
| NM_152913.2 | NP_690877.1 | 7 | 1234 | |||
| GenBank mRNA | AB072903.1 | - | 2 | 84 | ||
| AK124634.1 | - | 7 | 2111 | |||
| AK225783.1 | - | 7 | 1177 | |||
| AK294658.1 | - | 7 | 1050 | |||
| AK295543.1 | - | 6 | 892 | |||
| AK312364.1 | - | 7 | 1104 | |||
| AL832523.1 | - | 6 | 968 | |||
| AL834348.1 | - | 7 | 1080 | |||
| AL834349.1 | - | 7 | 1050 | |||
| AY358583.1 | - | 8 | 1166 | |||
| BC030793.1 | - | 7 | 1203 | 5263130 | ||

TMEM130
Genomic Map