siRNA Details
| Gene Symbol: NYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173564.3 | NP_775835.2 | 4 | 2075 | ||
| XM_006715907.3 | XP_006715970.1 | 4 | 2130 | |||
| XM_017011868.1 | XP_016867357.1 | 4 | 2130 | |||
| GenBank mRNA | AK094857.1 | - | 1 | 186 | ||
| AK124773.1 | - | 4 | 2075 | |||
| AY375450.1 | - | 1 | 164 | |||

NYAP1
Genomic Map