siRNA Details
| Gene Symbol: DEFB112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037498.1 | NP_001032587.1 | 2 | 182 | ||
| GenBank mRNA | AY122469.1 | - | 1 | 26 | ||
| DQ012016.1 | - | 2 | 182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DEFB112| Gene Symbol: DEFB112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037498.1 | NP_001032587.1 | 2 | 182 | ||
| GenBank mRNA | AY122469.1 | - | 1 | 26 | ||
| DQ012016.1 | - | 2 | 182 | |||