siRNA Details
| Gene Symbol: DEFB124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037500.1 | NP_001032589.1 | 2 | 127 | ||
| XM_017027782.1 | XP_016883271.1 | 3 | 744 | |||
| GenBank mRNA | AY122476.1 | - | 1 | 62 | ||
| DQ119827.1 | - | 2 | 127 | |||

DEFB124
Genomic Map