siRNA Details
| Gene Symbol: RNASEH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286834.1 | NP_001273763.1 | 7 | 822 | ||
| NM_001286837.1 | NP_001273766.1 | 7 | 1060 | |||
| NM_002936.4 | NP_002927.2 | 7 | 822 | |||
| GenBank mRNA | AF039652.1 | - | 7 | 792 | ||
| AF048994.1 | - | 7 | 738 | |||
| AF048995.1 | - | 7 | 772 | |||
| AJ224117.1 | - | 7 | 711 | |||
| AK075490.1 | - | 7 | 771 | |||
| AK096913.1 | - | 7 | 978 | |||
| BC002973.1 | - | 7 | 781 | 3537074 | ||
| BT019670.1 | - | 7 | 711 | |||
| CR541820.1 | - | 7 | 711 | |||

RNASEH1
Genomic Map