siRNA Details
| Gene Symbol: TMEM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288564.1 | NP_001275493.1 | 3 | 237 | ||
| NM_001288565.1 | NP_001275494.1 | 2 | 740 | |||
| NM_001288566.1 | NP_001275495.1 | 3 | 338 | |||
| NM_001288567.1 | NP_001275496.1 | 3 | 552 | |||
| NM_001288568.1 | NP_001275497.1 | 3 | 390 | |||
| NM_001288569.1 | NP_001275498.1 | 3 | 251 | |||
| NM_001288570.1 | NP_001275499.1 | 3 | 264 | |||
| NM_001288571.1 | NP_001275500.1 | 3 | 330 | |||
| NM_016456.4 | NP_057540.1 | 3 | 255 | |||
| NR_109920.1 | 3 | 255 | ||||
| NR_109921.1 | 2 | 740 | ||||
| XM_011509385.1 | XP_011507687.1 | 3 | 314 | |||
| XM_011509386.2 | XP_011507688.1 | 3 | 289 | |||
| XM_011509387.2 | XP_011507689.1 | 3 | 177 | |||
| GenBank mRNA | AB013909.1 | - | 2 | 108 | ||
| AF151020.1 | - | 4 | 227 | |||
| AK054883.1 | - | 3 | 210 | |||
| AK075335.1 | - | 3 | 165 | |||
| AK091726.1 | - | 2 | 625 | |||
| AK296307.1 | - | 3 | 172 | |||
| AK303843.1 | - | 3 | 276 | |||
| AL136600.1 | - | 3 | 188 | |||
| AL516746.3 | - | 3 | 430 | |||
| AY138587.1 | - | 3 | 166 | |||
| AY359012.1 | - | 3 | 229 | |||
| BC001106.2 | - | 3 | 179 | 3506442 | ||
| BE782946.1 | - | 3 | 217 | |||
| BM928361.1 | - | 3 | 177 | |||
| BX337142.2 | - | 2 | 624 | |||
| CR457162.1 | - | 2 | 108 | |||
| CR533467.1 | - | 2 | 108 | |||
| CX751563.1 | - | 2 | 622 | |||
| DA141558.1 | - | 3 | 237 | |||
| DA364005.1 | - | 3 | 255 | |||
| DB031447.1 | - | 3 | 231 | |||

TMEM9
Genomic Map